accessibility menu, dialog, popup

UserName

HONH2

CAS RN 23160-46-5

IUPAC Name: 10-aminodecan-1-ol
Synonyms: 10-aminodecan-1-ol
Molecular Weight (g/mol): 173.3
Molecular Formula: C10H23NO
InChi Key: SWVSKCPPMNGBGL-UHFFFAOYSA-N
SMILES: NCCCCCCCCCCO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results