missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-Amino-1-decanol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 72°C |
| Color | White-Yellow |
| Boiling Point | 154°C |
| Quantity | 5 g |
| Formula Weight | 173.30 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 10-Amino-1-decanol |
Safety and Handling
| TSCA | No |