missing translation for 'onlineSavingsMsg'
Learn More

10,15-Dihydro-5H-diindolo[3,2-a:3',2'-c]carbazole 98.0+%, TCI America™

Catalog No. D5428200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5428200MG 200 mg
D54281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5428200MG Supplier TCI America Supplier No. D5428200MG

Chemical Identifiers

CAS 109005-10-9
Molecular Formula C24H15N3
Molecular Weight (g/mol) 345.405
MDL Number MFCD21609440
InChI Key IQRFZFGTHZJRFV-UHFFFAOYSA-N
Synonym 10,15-Dihydro-5H-5,10,15-triazadiindeno[1,2-a:1′,2′-c]fluorene
PubChem CID 13707158
SMILES C1=CC=C2C(=C1)C3=C(N2)C4=C(C5=C3NC6=CC=CC=C65)NC7=CC=CC=C74

Specifications

Melting Point 350°C
Quantity 200 mg
Formula Weight 345.41
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 10,15-Dihydro-5H-diindolo[3,2-a:3′,2′-c]carbazole