accessibility menu, dialog, popup

UserName

CAS RN 109005-10-9

HNNHHN

CAS RN 109005-10-9

IUPAC Name: 9,18,27-triazaheptacyclo[18.7.0.0²,¹⁰.0³,⁸.0¹¹,¹⁹.0¹²,¹⁷.0²¹,²⁶]heptacosa-1(20),2(10),3,5,7,11(19),12,14,16,21(26),22,24-dodecaene
Synonyms: 9,18,27-triazaheptacyclo[18.7.0.0²,¹⁰.0³,⁸.0¹¹,¹⁹.0¹²,¹⁷.0²¹,²⁶]heptacosa-1(20),2(10),3,5,7,11(19),12,14,16,21(26),22,24-dodecaene
Molecular Weight (g/mol): 345.41
Molecular Formula: C24H15N3
InChi Key: IQRFZFGTHZJRFV-UHFFFAOYSA-N
SMILES: N1C2=CC=CC=C2C2=C1C1=C(NC3=CC=CC=C13)C1=C2NC2=C1C=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results