missing translation for 'onlineSavingsMsg'
Learn More

10,12-Pentacosadiynoic acid, 98+%

CAS: 66990-32-7 | C25H42O2 | 374.61 g/mol

Supplier:  Thermo Scientific Chemicals L0931306

 View more versions of this product

Catalog No. AAL0931306

Only null left
Explore available promotions

Description

Description

10,12-Pentacosadiynoic acid is used in the preparation of colorimetric and fluorescence sensors for cationic surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10,12-Pentacosadiynoic acid is used in the preparation of colorimetric and fluorescence sensors for cationic surfactants.

Solubility
Slightly soluble in water.

Notes
Incompatible with reducing agents and oxidizing agents.
Specifications

Specifications

10,12-Pentacosadiynoic acid
62°C to 65°C
MFCD00041684
4189973
10,12-pentacosadiynoic acid, 10,12-pentacosadiynoicacid, acmc-209nx9, 10-12-pentacosadiynoic acid, 10,12-pentacosadiynoic acid hplc
ZPUDRBWHCWYMQS-UHFFFAOYSA-N
pentacosa-10,12-diynoic acid
538433
≥98%
66990-32-7
C25H42O2
5 g
Light sensitive
Slightly soluble in water.
CCCCCCCCCCCCC#CC#CCCCCCCCCC(O)=O
374.61
374.61

Chemical Identifiers

66990-32-7
374.61
ZPUDRBWHCWYMQS-UHFFFAOYSA-N
538433
CCCCCCCCCCCCC#CC#CCCCCCCCCC(O)=O
C25H42O2
MFCD00041684
10,12-pentacosadiynoic acid, 10,12-pentacosadiynoicacid, acmc-209nx9, 10-12-pentacosadiynoic acid, 10,12-pentacosadiynoic acid hplc
pentacosa-10,12-diynoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 614-006-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only