missing translation for 'onlineSavingsMsg'
Learn More

10,12-Pentacosadiynoic acid, 98+%

Catalog No. p-7045064
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0931303 1 g
AAL0931306 5 g
AAL0931314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0931303 Supplier Thermo Scientific Chemicals Supplier No. L0931303
Only null left

CAS: 66990-32-7 | C25H42O2 | 374.61 g/mol

10,12-Pentacosadiynoic acid is used in the preparation of colorimetric and fluorescence sensors for cationic surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
10,12-Pentacosadiynoic acid is used in the preparation of colorimetric and fluorescence sensors for cationic surfactants.

Solubility
Slightly soluble in water.

Notes
Incompatible with reducing agents and oxidizing agents.

Chemical Identifiers

CAS 66990-32-7
Molecular Formula C25H42O2
Molecular Weight (g/mol) 374.61
MDL Number MFCD00041684
InChI Key ZPUDRBWHCWYMQS-UHFFFAOYSA-N
Synonym 10,12-pentacosadiynoic acid, 10,12-pentacosadiynoicacid, acmc-209nx9, 10-12-pentacosadiynoic acid, 10,12-pentacosadiynoic acid hplc
PubChem CID 538433
IUPAC Name pentacosa-10,12-diynoic acid
SMILES CCCCCCCCCCCCC#CC#CCCCCCCCCC(O)=O

Specifications

Melting Point 62°C to 65°C
Quantity 1 g
Beilstein 4189973
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 374.61
Percent Purity ≥98%
Chemical Name or Material 10,12-Pentacosadiynoic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 614-006-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only