missing translation for 'onlineSavingsMsg'
Learn More

10,12-Pentacosadiyn-1-ol 98.0+%, TCI America™

Catalog No. p-7134920
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1053100MG 100 mg
P10531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1053100MG Supplier TCI America Supplier No. P1053100MG
Only null left

Chemical Identifiers

CAS 92266-90-5
Molecular Formula C25H44O
Molecular Weight (g/mol) 360.626
MDL Number MFCD00059096
InChI Key XHKKFKTVKSZQKA-UHFFFAOYSA-N
PubChem CID 5122328
IUPAC Name pentacosa-10,12-diyn-1-ol
SMILES CCCCCCCCCCCCC#CC#CCCCCCCCCCO

Specifications

Melting Point 60°C
Color White
Quantity 100 mg
Formula Weight 360.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 10,12-Pentacosadiyn-1-ol