accessibility menu, dialog, popup

UserName

CAS RN 92266-90-5

H3COH

CAS RN 92266-90-5

IUPAC Name: pentacosa-10,12-diyn-1-ol
Synonyms: pentacosa-10,12-diyn-1-ol
Molecular Weight (g/mol): 360.63
Molecular Formula: C25H44O
InChi Key: XHKKFKTVKSZQKA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC#CC#CCCCCCCCCCO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results