missing translation for 'onlineSavingsMsg'
Learn More

10,12-Docosadiynedioic acid, 95%

Catalog No. AAL1064706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1064706 5 g
AAL1064703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1064706 Supplier Thermo Scientific Chemicals Supplier No. L1064706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 28393-02-4 | C22H34O4 | 362.51 g/mol

Polymerizable diacid (10,12-docosadiynedioic acid) was used as obtained from GFS Chemicals. Synthesis of dimesogenic liquid crystals with both cholesterol and azobenzene groups is by the esterification of 10,12-docosadiynedioic acid. .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Polymerizable diacid (10,12-docosadiynedioic acid) was used as obtained from GFS Chemicals. Synthesis of dimesogenic liquid crystals with both cholesterol and azobenzene groups is by the esterification of 10,12-docosadiynedioic acid. .

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.

Chemical Identifiers

CAS 28393-02-4
Molecular Formula C22H34O4
Molecular Weight (g/mol) 362.51
MDL Number MFCD00078306
InChI Key XCUAGNLOPZWTEH-UHFFFAOYSA-N
Synonym 10,12-docosadiynedioic acid, 10,12-docosadiyndioic acid, 10,12-docosadiynedioicacid, acmc-20aozt, 10,12-docasadiyndioic acid, 10,12-docosadiynedioic acid #, icosa-9,11-diyne-1,20-dicarboxylic acid
PubChem CID 544138
IUPAC Name docosa-10,12-diynedioic acid
SMILES C(CCCCC(=O)O)CCCC#CC#CCCCCCCCCC(=O)O

Specifications

Melting Point 110°C to 112°C
Quantity 5 g
Beilstein 1803395
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 362.51
Percent Purity 95%
Chemical Name or Material 10,12-Docosadiynedioic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only