CAS RN 28393-02-4
IUPAC Name:
docosa-10,12-diynedioic acid
Synonyms:
docosa-10,12-diynedioic acid
Molecular Weight (g/mol):
362.51
Molecular Formula:
C22H34O4
InChi Key:
XCUAGNLOPZWTEH-UHFFFAOYSA-N
SMILES:
OC(=O)CCCCCCCCC#CC#CCCCCCCCCC(O)=O
1
–
1
of
1
results
1
Promotions Available
10,12-Docosadiynedioic acid, 95%
10,12-Docosadiynedioic acid, 95%
CAS: 28393-02-4 Molecular Formula: C22H34O4 Molecular Weight (g/mol): 362.51 MDL Number: MFCD00078306 InChI Key: XCUAGNLOPZWTEH-UHFFFAOYSA-N Synonym: 10,12-docosadiynedioic acid,10,12-docosadiyndioic acid,10,12-docosadiynedioicacid,acmc-20aozt,10,12-docasadiyndioic acid,10,12-docosadiynedioic acid #,icosa-9,11-diyne-1,20-dicarboxylic acid PubChem CID: 544138 IUPAC Name: docosa-10,12-diynedioic acid SMILES: C(CCCCC(=O)O)CCCC#CC#CCCCCCCCCC(=O)O