Learn More
Agilent BioTek 1 x 16 Tube Syringe Dispenser Manifold
Catalog No. BT7180543S
Change view
Click to view available options
For Use With (Equipment):
EL406 Microplate Washer
Catalog No. For Use With (Equipment)
BT7180543S EL406 Microplate Washer
1 options
Catalog No. BT7180543S Supplier Agilent BioTek Supplier No. 7180543S
Dispenser manifold for use with the MultiFlo FX, EL406 and 406 FX for faster 96- and 384-well microplate dispensing.
  • Autoclavable
  • Straight, steel tips
  • 2 are required for Dual Syringe Dispenser
Specifications
Type Manifold
For Use With (Equipment) EL406 Microplate Washer