missing translation for 'onlineSavingsMsg'
Learn More

1-Vinylimidazole 98.0+%, TCI America™

Catalog No. V004525ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V004525ML 25 mL
V0045100ML 100 mL
V0045500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. V004525ML Supplier TCI America Supplier No. V004525ML

Chemical Identifiers

CAS 1072-63-5
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.12
MDL Number MFCD00005297
InChI Key OSSNTDFYBPYIEC-UHFFFAOYSA-N
Synonym 1-vinylimidazole, 1-vinyl-1h-imidazole, n-vinylimidazole, 1h-imidazole, 1-ethenyl, lufixan, polyvinylimidazole, poly vinylimidazole, poly n-vinylimidazole, poly 1-vinylimidazole, n-vinylimidazole polymer
PubChem CID 66171
IUPAC Name 1-ethenyl-1H-imidazole
SMILES C=CN1C=CN=C1

Specifications

Color Yellow
pH 9 to 11 (100g/L 25°C)
Boiling Point 80°C
Quantity 25 mL
UN Number 3267
Formula Weight 94.12
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Vinylimidazole
EINECSNumber (5)-5990
TSCA Yes