accessibility menu, dialog, popup

UserName

H2CNN

CAS RN 1072-63-5

IUPAC Name: 1-ethenyl-1H-imidazole
Synonyms: 1-ethenylimidazole
Molecular Weight (g/mol): 94.12
Molecular Formula: C5H6N2
InChi Key: OSSNTDFYBPYIEC-UHFFFAOYSA-N
SMILES: C=CN1C=CN=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)