missing translation for 'onlineSavingsMsg'
Learn More

1-Vinylimidazole, 99%

Catalog No. AAL1617414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1617414 25 g
AAL1617422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1617414 Supplier Thermo Scientific Chemicals Supplier No. L1617414

CAS: 1072-63-5 | C5H6N2 | 94.12 g/mol

1-Vinylimidazole is used as a monomer in the synthesis of molecularly imprinted polymers. It is involved in the removal of vinyl protecting groups with an isoprene-catalyzed lithiation process. Also, it is involved in the preparation of magnetic-poly(divinylbenzene-1-vinylimidazole) microbeads. Further, it is used to prepare ion imprinted polyvinylimidazole-silica hybrid copolymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Vinylimidazole is used as a monomer in the synthesis of molecularly imprinted polymers. It is involved in the removal of vinyl protecting groups with an isoprene-catalyzed lithiation process. Also, it is involved in the preparation of magnetic-poly(divinylbenzene-1-vinylimidazole) microbeads. Further, it is used to prepare ion imprinted polyvinylimidazole-silica hybrid copolymer.

Solubility
Miscible with water.

Notes
Hygroscopic. Light sensitive. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 1072-63-5
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.12
MDL Number MFCD00005297
InChI Key OSSNTDFYBPYIEC-UHFFFAOYSA-N
Synonym 1-vinylimidazole, 1-vinyl-1h-imidazole, n-vinylimidazole, 1h-imidazole, 1-ethenyl, lufixan, polyvinylimidazole, poly vinylimidazole, poly n-vinylimidazole, poly 1-vinylimidazole, n-vinylimidazole polymer
PubChem CID 66171
SMILES C=CN1C=CN=C1

Specifications

Melting Point <-50°C
Density 1.038
Boiling Point 192°C to 194°C
Flash Point 84°C (183°F)
Refractive Index 1.5315
Quantity 25 g
UN Number UN3267
Beilstein 105321
Sensitivity Hygroscopic
Solubility Information Miscible with water.
IUPAC Name 1-ethenylimidazole
Formula Weight 94.12
Percent Purity 99%
Chemical Name or Material 1-Vinylimidazole
Show More Show Less
Hazard Category H227-H302-H318-H360D
Hazard Statement GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P201-P202-P210-P264b-P270-P281-P301+P312-P305+P351+P338-P308+P313-P310-P330-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 214-012-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only