Learn More
1-Undecene, 97%
CAS: 821-95-4 | C11H22 | 154.297 g/mol
$118.44 - $496.05
Chemical Identifiers
| CAS | 821-95-4 |
|---|---|
| Molecular Formula | C11H22 |
| Molecular Weight (g/mol) | 154.297 |
| MDL Number | MFCD00008956 |
| InChI Key | DCTOHCCUXLBQMS-UHFFFAOYSA-N |
| Synonym | 1-undecene, undecene, n-1-undecene, alpha-undecene, alpha-undecylene, 1-hendecene, alpha-nonylethylene, undecene-1, undecene petroleum, ccris 5720 |
| PubChem CID | 13190 |
| ChEBI | CHEBI:77444 |
| IUPAC Name | undec-1-ene |
| SMILES | CCCCCCCCCC=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5338706
|
Thermo Scientific Chemicals
H5338706 |
5 g |
Each for $118.44
|
|
|||||
|
AAH5338714
|
Thermo Scientific Chemicals
H5338714 |
25 g |
Each for $496.05
|
|
|||||
Description
1-Undecene was used as a biomarker for identification of Pseudomonas aeruginosa strain. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Undecene was used as a biomarker for identification of Pseudomonas aeruginosa strain. It is also used as pharmaceutical intermediate.
Solubility
Solunle in ether, chloroform and ligroin. Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 821-95-4 | |
| 154.297 | |
| DCTOHCCUXLBQMS-UHFFFAOYSA-N | |
| 13190 | |
| undec-1-ene |
| C11H22 | |
| MFCD00008956 | |
| 1-undecene, undecene, n-1-undecene, alpha-undecene, alpha-undecylene, 1-hendecene, alpha-nonylethylene, undecene-1, undecene petroleum, ccris 5720 | |
| CHEBI:77444 | |
| CCCCCCCCCC=C |
Specifications
| 821-95-4 | |
| 0.751 | |
| 63°C (145°F) | |
| 1.426 | |
| MFCD00008956 | |
| 1740044 | |
| Solunle in ether,chloroform and ligroin. Insoluble in water. | |
| CCCCCCCCCC=C | |
| 154.297 | |
| CHEBI:77444 | |
| 97% |
| -49°C | |
| 192°C to 193°C | |
| C11H22 | |
| CH3(CH2)8CH=CH2 | |
| 5 g | |
| 1-undecene, undecene, n-1-undecene, alpha-undecene, alpha-undecylene, 1-hendecene, alpha-nonylethylene, undecene-1, undecene petroleum, ccris 5720 | |
| DCTOHCCUXLBQMS-UHFFFAOYSA-N | |
| undec-1-ene | |
| 13190 | |
| 154.3 | |
| 1-Undecene |
Safety and Handling
GHS H Statement
H302-H332-H315-H319-H335-H227
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H332-H315-H319-H335
EINECSNumber : 212-483-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only