missing translation for 'onlineSavingsMsg'
Learn More

1-Undecanol 99.0+%, TCI America™

Catalog No. p-7137834
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U000525ML 25 mL
U0005100ML 100 mL
U0005500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. U000525ML Supplier TCI America Supplier No. U000525ML
Only null left

Chemical Identifiers

CAS 112-42-5
Molecular Formula C11H24O
Molecular Weight (g/mol) 172.312
MDL Number MFCD00004751
InChI Key KJIOQYGWTQBHNH-UHFFFAOYSA-N
Synonym 1-undecanol, undecyl alcohol, undecanol, n-undecanol, n-undecyl alcohol, hendecanoic alcohol, hendecyl alcohol, 1-hendecanol, n-hendecylenic alcohol, tip-nip
PubChem CID 8184
ChEBI CHEBI:87499
IUPAC Name undecan-1-ol
SMILES CCCCCCCCCCCO

Specifications

Color Yellow
Boiling Point 147°C
Quantity 25 mL
UN Number 3082
Formula Weight 172.31
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Undecanol
EINECSNumber (2)-0217&(2)-3704
RTECSNumber YQ3155000
TSCA Yes