accessibility menu, dialog, popup

UserName

H3COH

CAS RN 112-42-5

IUPAC Name: undecan-1-ol
Synonyms: undecanol
Molecular Weight (g/mol): 172.31
Molecular Formula: C11H24O
InChi Key: KJIOQYGWTQBHNH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCO
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)