missing translation for 'onlineSavingsMsg'
Learn More

1-Tritylimidazole-4-carboxaldehyde 98.0+%, TCI America™

Catalog No. T22041G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22041G 1 g
T2204100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22041G Supplier TCI America Supplier No. T22041G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33016-47-6
Molecular Formula C23H18N2O
Molecular Weight (g/mol) 338.41
MDL Number MFCD02179554
InChI Key YQYLLBSWWRWWAY-UHFFFAOYSA-N
Synonym 1-tritylimidazole-4-carboxaldehyde, 1-trityl-1h-imidazole-4-carbaldehyde, 1-trityl-1h-imidazole-4-carboxaldehyde, 1-trityl-imdazole-4-carboxadehyde, 4-formyl-1-tritylimidazole, 4-formyl-1-trityl-1h-imidazole, 1-trityl-4-formylimidazole, 1-triphenylmethyl imidazole-4-carbaldehyde, 1-triphenylmethyl-1h-imidazole-4-carbaldehyde, 1h-imidazole-4-carboxaldehyde, 1-triphenylmethyl
PubChem CID 618233
IUPAC Name 1-(triphenylmethyl)-1H-imidazole-4-carbaldehyde
SMILES O=CC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 186°C
Quantity 1 g
Formula Weight 338.41
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Tritylimidazole-4-carboxaldehyde