missing translation for 'onlineSavingsMsg'
Learn More

1-Tritylimidazole-4-carboxaldehyde, 98%

Catalog No. p-7028131
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2527206 5 g
AAB2527214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2527206 Supplier Thermo Scientific Chemicals Supplier No. B2527206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33016-47-6 | C23H18N2O | 338.41 g/mol

1-Tritylimidazole-4-carboxaldehyde is used in the synthesis of histamine and girolline

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Tritylimidazole-4-carboxaldehyde is used in the synthesis of histamine and girolline

Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agents, air.

Chemical Identifiers

CAS 33016-47-6
Molecular Formula C23H18N2O
Molecular Weight (g/mol) 338.41
MDL Number MFCD02179554
InChI Key YQYLLBSWWRWWAY-UHFFFAOYSA-N
Synonym 1-tritylimidazole-4-carboxaldehyde, 1-trityl-1h-imidazole-4-carbaldehyde, 1-trityl-1h-imidazole-4-carboxaldehyde, 1-trityl-imdazole-4-carboxadehyde, 4-formyl-1-tritylimidazole, 4-formyl-1-trityl-1h-imidazole, 1-trityl-4-formylimidazole, 1-triphenylmethyl imidazole-4-carbaldehyde, 1-triphenylmethyl-1h-imidazole-4-carbaldehyde, 1h-imidazole-4-carboxaldehyde, 1-triphenylmethyl
PubChem CID 618233
SMILES O=CC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point ∼192°C (decomposition)
Quantity 5 g
Sensitivity Air Sensitive
IUPAC Name 1-tritylimidazole-4-carbaldehyde
Formula Weight 338.41
Percent Purity 98%
Chemical Name or Material 1-Tritylimidazole-4-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only