missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Trimethylsilyl)-1H-benzotriazole 97.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 5 g |
| UN Number | 1993 |
| Formula Weight | 191.31 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-(Trimethylsilyl)-1H-benzotriazole |
Safety and Handling
| TSCA | No |