CAS RN 43183-36-4
IUPAC Name:
1-(trimethylsilyl)-1H-1,2,3-benzotriazole
Synonyms:
1-(trimethylsilyl)-1,2,3-benzotriazole
Molecular Weight (g/mol):
191.31
Molecular Formula:
C9H13N3Si
InChi Key:
VHOLOZYRMQVHBK-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)N1N=NC2=CC=CC=C12
1
–
1
of
1
results
1
1-(Trimethylsilyl)-1H-benzotriazole 97.0+%, TCI America™
CAS: 43183-36-4 Molecular Formula: C9H13N3Si Molecular Weight (g/mol): 191.309 MDL Number: MFCD00216577 InChI Key: VHOLOZYRMQVHBK-UHFFFAOYSA-N Synonym: 1-trimethylsilyl-1h-benzotriazole,1-trimethylsilylbenzotriazole,1-trimethylsilyl-1h-benzo d 1,2,3 triazole,1-trimethylsilyl-1,2,3-benzotriazole,1-tms-1h-benzotriazole,acmc-1bn8x,n-trimethylsilylbenzotriazole,benzotriazol-1-yl trimethyl silane,1-trimethylsilanyl-1h-benzotriazole PubChem CID: 3558326 IUPAC Name: benzotriazol-1-yl(trimethyl)silane SMILES: C[Si](C)(C)N1C2=CC=CC=C2N=N1