missing translation for 'onlineSavingsMsg'
Learn More

1-(Trimethylsilyl)-1-propyne 98.0+%, TCI America™

Catalog No. p-7136620
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11235ML 5 mL
T112325ML 25 mL
T1123100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T11235ML Supplier TCI America Supplier No. T11235ML
Only null left

Chemical Identifiers

CAS 6224-91-5
Molecular Formula C6H12Si
Molecular Weight (g/mol) 112.247
MDL Number MFCD00009271
InChI Key DCGLONGLPGISNX-UHFFFAOYSA-N
Synonym 1-trimethylsilyl-1-propyne, 1-trimethylsilyl propyne, trimethyl prop-1-yn-1-yl silane, 1-trimethylsilylpropyne, 1-trimethylsilyl prop-1-yne, trimethylsilylpropyne, trimethyl prop-1-ynyl silane, silane, trimethyl-1-propynyl, trimethyl-prop-1-ynyl-silane
PubChem CID 80363
IUPAC Name trimethyl(prop-1-ynyl)silane
SMILES CC#C[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 100°C
Quantity 5 mL
UN Number 1993
Formula Weight 112.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(Trimethylsilyl)-1-propyne
TSCA No
Recommended Storage Refrigerator