accessibility menu, dialog, popup

UserName

CAS RN 6224-91-5

IUPAC Name: trimethyl(prop-1-yn-1-yl)silane
Synonyms: trimethyl(prop-1-yn-1-yl)silane
Molecular Weight (g/mol): 112.25
Molecular Formula: C6H12Si
InChi Key: DCGLONGLPGISNX-UHFFFAOYSA-N
SMILES: CC#C[Si](C)(C)C
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)