missing translation for 'onlineSavingsMsg'
Learn More

1-(Trimethylsilyl)-1-pentyne 98.0+%, TCI America™

Catalog No. T27465G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T27465G Supplier TCI America Supplier No. T27465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18270-17-2
Molecular Formula C8H16Si
Molecular Weight (g/mol) 140.30
MDL Number MFCD00077890
InChI Key CABCDUQQPBAHEE-UHFFFAOYSA-N
Synonym 1-trimethylsilyl-1-pentyne, trimethyl pent-1-yn-1-yl silane, trimethyl pent-1-ynyl silane, 1-pentyn-1-yltrimethylsilane, 1-tms-1-pentyne, acmc-209ejs, 1-trimethylsilylpent-1-yne, 1-trimethylsi;lyl-1-pentyne
PubChem CID 2760860
IUPAC Name trimethyl(pent-1-yn-1-yl)silane
SMILES CCCC#C[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 g
UN Number 1993
Formula Weight 140.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(Trimethylsilyl)-1-pentyne