accessibility menu, dialog, popup

UserName

CAS RN 18270-17-2

IUPAC Name: trimethyl(pent-1-yn-1-yl)silane
Synonyms: trimethyl(pent-1-yn-1-yl)silane
Molecular Weight (g/mol): 140.3
Molecular Formula: C8H16Si
InChi Key: CABCDUQQPBAHEE-UHFFFAOYSA-N
SMILES: CCCC#C[Si](C)(C)C
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results