missing translation for 'onlineSavingsMsg'
Learn More

1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole 97.0+%, TCI America™

Catalog No. T26241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26241G 1 g
T26245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26241G Supplier TCI America Supplier No. T26241G

Chemical Identifiers

CAS 887144-97-0
Molecular Formula C10H10F3IO
Molecular Weight (g/mol) 330.089
MDL Number MFCD10567056
InChI Key HVAPLSNCVYXFDQ-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole, 1-trifluoromethyl-3,3-dimethyl-1,2-benziodoxole, togni's reagent, togni reagent, unii-3ok0e02mt6, 1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl, 1,3-dihydro-3,3-dimethyl-1-trifluoromethyl-1,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl-1,3-dihydro-1lambda3,2-benziodoxole, 3,3-dimethyl-1-trifluoromethyl-1? 3 ,2-benziodaoxole, togni's reagents dimethyl compound mi
PubChem CID 16043572
IUPAC Name 3,3-dimethyl-1-(trifluoromethyl)-1$l^{3},2-benziodoxole
SMILES CC1(C2=CC=CC=C2I(O1)C(F)(F)F)C

Specifications

Melting Point 78°C
Color White-Yellow
Quantity 1 g
Formula Weight 330.09
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole
TSCA No
Recommended Storage Refrigerator