missing translation for 'onlineSavingsMsg'
Learn More

1-(Trifluoroacetyl)piperidine 98.0+%, TCI America™

Catalog No. T20875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20875G 5 g
T208725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20875G Supplier TCI America Supplier No. T20875G
Only null left

Chemical Identifiers

CAS 340-07-8
Molecular Formula C7H10F3NO
Molecular Weight (g/mol) 181.158
MDL Number MFCD00466213
InChI Key BCJUMGSHTLYECD-UHFFFAOYSA-N
PubChem CID 5181830
IUPAC Name 2,2,2-trifluoro-1-piperidin-1-ylethanone
SMILES C1CCN(CC1)C(=O)C(F)(F)F

Specifications

Color Colorless
Boiling Point 77°C
Quantity 5 g
Formula Weight 181.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(Trifluoroacetyl)piperidine