missing translation for 'onlineSavingsMsg'
Learn More

1-Tridecene, 97%

CAS: 2437-56-1 | C13H26 | 182.351 g/mol

$91.63 - $273.62

Chemical Identifiers

CAS 2437-56-1
Molecular Formula C13H26
Molecular Weight (g/mol) 182.351
MDL Number MFCD00008976
InChI Key VQOXUMQBYILCKR-UHFFFAOYSA-N
Synonym 1-tridecene, n-tridec-1-ene, tridecylene, alpha-tridecene, undecylethylene, tridecene-1, unii-5b0u2s314c, ccris 5719, 1-tridecylene, .alpha.-tridecene
PubChem CID 17095
IUPAC Name tridec-1-ene
SMILES CCCCCCCCCCCC=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2441306
View Documents
Thermo Scientific Chemicals
B2441306
5 g
Each for $91.63
Only null left
 
AAB2441314
View Documents
Thermo Scientific Chemicals
B2441314
25 g
Each for $273.62
Only null left
 
Description

Description

It is used in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

2437-56-1
182.351
VQOXUMQBYILCKR-UHFFFAOYSA-N
17095
CCCCCCCCCCCC=C
C13H26
MFCD00008976
1-tridecene, n-tridec-1-ene, tridecylene, alpha-tridecene, undecylethylene, tridecene-1, unii-5b0u2s314c, ccris 5719, 1-tridecylene, .alpha.-tridecene
tridec-1-ene

Specifications

2437-56-1
0.766
79°C (174°F)
C13H26
CH3(CH2)10CH=CH2
5 g
1-tridecene, n-tridec-1-ene, tridecylene, alpha-tridecene, undecylethylene, tridecene-1, unii-5b0u2s314c, ccris 5719, 1-tridecylene, .alpha.-tridecene
VQOXUMQBYILCKR-UHFFFAOYSA-N
tridec-1-ene
17095
97%
-23°C
232°C to 233°C
Mild
1.433
MFCD00008976
1744660
Insoluble in water.
CCCCCCCCCCCC=C
182.351
182.35
1-Tridecene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 219-443-8

RTECSNumber : YD4595000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only