missing translation for 'onlineSavingsMsg'
Learn More

1-Tridecanol 98.0+%, TCI America™

Catalog No. T080325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T080325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T080325G Supplier TCI America Supplier No. T080325G

Chemical Identifiers

CAS 112-70-9
Molecular Formula C13H28O
Molecular Weight (g/mol) 200.366
MDL Number MFCD00004756
InChI Key XFRVVPUIAFSTFO-UHFFFAOYSA-N
Synonym 1-tridecanol, tridecanol, tridecyl alcohol, n-tridecan-1-ol, n-tridecanol, n-tridecyl alcohol, alcohols, c12-14, ccris 8591, dsstox_cid_1947, dsstox_rid_76420
PubChem CID 8207
ChEBI CHEBI:34123
IUPAC Name tridecan-1-ol
SMILES CCCCCCCCCCCCCO

Specifications

Color White
Quantity 25 g
Formula Weight 200.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Tridecanol
EINECSNumber (2)-0217&(2)-3704
RTECSNumber YD4200000
TSCA Yes