missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Triacontanol 85.0+%, TCI America™
$296.48 - $1628.69
Chemical Identifiers
| CAS | 593-50-0 |
|---|---|
| Molecular Formula | C30H62O |
| Molecular Weight (g/mol) | 438.825 |
| MDL Number | MFCD00002963 |
| InChI Key | REZQBEBOWJAQKS-UHFFFAOYSA-N |
| Synonym | 1-triacontanol, triacontanol, melissyl alcohol, myricyl alcohol, n-triacontanol, triacontyl alcohol, triacontanol-1, miraculan, myricyl alcohol van, 1-hydroxytriacontane |
| PubChem CID | 68972 |
| ChEBI | CHEBI:28409 |
| IUPAC Name | triacontan-1-ol |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T1049100MG
|
TCI America
T1049100MG |
100 mg |
Each for $296.48
|
|
|||||
|
T10491G
|
TCI America
T10491G |
1 g |
Each for $1,628.69
|
|
|||||
Chemical Identifiers
| 593-50-0 | |
| 438.825 | |
| REZQBEBOWJAQKS-UHFFFAOYSA-N | |
| 68972 | |
| triacontan-1-ol |
| C30H62O | |
| MFCD00002963 | |
| 1-triacontanol, triacontanol, melissyl alcohol, myricyl alcohol, n-triacontanol, triacontyl alcohol, triacontanol-1, miraculan, myricyl alcohol van, 1-hydroxytriacontane | |
| CHEBI:28409 | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCO |
Specifications
| 593-50-0 | |
| C30H62O | |
| 100 mg | |
| REZQBEBOWJAQKS-UHFFFAOYSA-N | |
| triacontan-1-ol | |
| 68972 | |
| 438.83 | |
| Crystalline Powder |
| 86°C | |
| MFCD00002963 | |
| 1-triacontanol, triacontanol, melissyl alcohol, myricyl alcohol, n-triacontanol, triacontyl alcohol, triacontanol-1, miraculan, myricyl alcohol van, 1-hydroxytriacontane | |
| CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCO | |
| 438.825 | |
| CHEBI:28409 | |
| ≥85.0% (GC) | |
| 1-Triacontanol |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : Yes