accessibility menu, dialog, popup

UserName

CAS RN 593-50-0

IUPAC Name: triacontan-1-ol
Synonyms: triacontanol aluminum salt 1-triacontanol myricyl alcohol melissyl alcohol
Molecular Weight (g/mol): 438.83
Molecular Formula: C30H62O
InChi Key: REZQBEBOWJAQKS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCO
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)