missing translation for 'onlineSavingsMsg'
Learn More

1-Triacontanol 85.0+%, TCI America™

Catalog No. T1049100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1049100MG 100 mg
T10491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1049100MG Supplier TCI America Supplier No. T1049100MG

Chemical Identifiers

CAS 593-50-0
Molecular Formula C30H62O
Molecular Weight (g/mol) 438.825
MDL Number MFCD00002963
InChI Key REZQBEBOWJAQKS-UHFFFAOYSA-N
Synonym 1-triacontanol, triacontanol, melissyl alcohol, myricyl alcohol, n-triacontanol, triacontyl alcohol, triacontanol-1, miraculan, myricyl alcohol van, 1-hydroxytriacontane
PubChem CID 68972
ChEBI CHEBI:28409
IUPAC Name triacontan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCO

Specifications

Melting Point 86°C
Quantity 100 mg
Formula Weight 438.83
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Triacontanol
EINECSNumber (2)-0217
TSCA Yes