missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Triacontanol, 96%
CAS: 593-50-0 | C30H62O | 438.82 g/mol
Supplier: Thermo Scientific Chemicals 218131000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Triacontanol | |
593-50-0 | |
100.0 | |
Authentic | |
Glass bottle | |
CH3(CH2)29OH | |
100 mg | |
1-triacontanol, triacontanol, melissyl alcohol, myricyl alcohol, n-triacontanol, triacontyl alcohol, triacontanol-1, miraculan, myricyl alcohol van, 1-hydroxytriacontane | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCO | |
438.82 | |
CHEBI:28409 | |
96% |
96% | |
95.0 | |
Beige | |
96% | |
C30H62O | |
MFCD00002963 | |
15, 9753 | |
REZQBEBOWJAQKS-UHFFFAOYSA-N | |
triacontan-1-ol | |
68972 | |
438.82 | |
Flakes |
Chemical Identifiers
593-50-0 | |
438.82 | |
REZQBEBOWJAQKS-UHFFFAOYSA-N | |
68972 | |
triacontan-1-ol |
C30H62O | |
MFCD00002963 | |
1-triacontanol, triacontanol, melissyl alcohol, myricyl alcohol, n-triacontanol, triacontyl alcohol, triacontanol-1, miraculan, myricyl alcohol van, 1-hydroxytriacontaneShow More | |
CHEBI:28409 | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCO |
Safety and Handling
EINECSNumber : 209-794-5
RUO – Research Use Only