missing translation for 'onlineSavingsMsg'
Learn More

1-Thionaphthol, 99%

Catalog No. p-7027838
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2487603 1 g
AAB2487606 5 g
AAB2487614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2487603 Supplier Thermo Scientific Chemicals Supplier No. B2487603
Only null left

CAS: 529-36-2 | C10H7S | 159.23 g/mol

1-Thionaphthol is used to produce the di-[1]naphthyl disulfide. It is also used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Thionaphthol is used to produce the di-[1]naphthyl disulfide. It is also used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in diethyl ether and ethanol. Slightly soluble in water and dilute alkalis.

Notes
Store away from air. Store under dry inert gas. Incompatible with air and oxidizing agents.

Chemical Identifiers

CAS 529-36-2
Molecular Formula C10H7S
Molecular Weight (g/mol) 159.23
MDL Number MFCD00039599
InChI Key SEXOVMIIVBKGGM-UHFFFAOYSA-M
Synonym 1-naphthalenethiol, 1-thionaphthol, naphthalenethiol, 1-naphthylthiol, unii-9a78cp495h, 1-mercaptonaphthalene, .alpha.-naphthyl mercaptan, naphthalene thiol, 1-napthalenethiol, naphthalen-1-thiol
PubChem CID 68259
IUPAC Name naphthalene-1-thiol
SMILES [S-]C1=C2C=CC=CC2=CC=C1

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in diethyl ether and ethanol. Slightly soluble in water and dilute alkalis.
Formula Weight 160.24
Percent Purity 99%
Chemical Name or Material 1-Thionaphthol
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 208-462-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only