missing translation for 'onlineSavingsMsg'
Learn More

1-Thio-beta-D-glucose tetraacetate, 98+%

Catalog No. aal1140403
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
Catalog No. Quantity
AAL1140403 1 g
AAL11404MD 250 mg
2 options

Catalog No. AAL1140403

Supplier: Thermo Scientific Chemicals L1140403

Only null left

CAS: 19879-84-6 | C14H20O9S | 364.37 g/mol

1-Thio-beta-D-glucose tetraacetate is used as an inhibitor of the Maillard reaction between glucose and glycine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Thio-beta-D-glucose tetraacetate is used as an inhibitor of the Maillard reaction between glucose and glycine.

Solubility
Soluble in water and chloroform (50 mg/ml).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 19879-84-6
Molecular Formula C14H20O9S
Molecular Weight (g/mol) 364.37
MDL Number MFCD00063271
InChI Key SFOZKJGZNOBSHF-UHFFFAOYNA-N
Synonym 1-thio-beta-d-glucose tetraacetate, 2r,3r,4s,5r,6s-2-acetoxymethyl-6-mercaptotetrahydro-2h-pyran-3,4,5-triyl triacetate, 1-thio-b-d-glucose tetraacetate, 1-thio-beta-d-glucose 2,3,4,6-tetraacetate, 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-glucopyranose, 1-thio-, a-d-glucose tetraacetate, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-sulfanyloxan-2-yl methyl acetate, 2,3,4,6-tetra-o-acetyl-1-thio-, a-d-glucopyranose, beta-d-thioglucose tetraacetate, 1-thio-ss-d-glucose tetraacetate
PubChem CID 88293
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-sulfanyloxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(S)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 115°C to 117°C
Quantity 1 g
Beilstein 1440689
Solubility Information Soluble in water and chloroform (50mg/ml).
Optical Rotation +5.5° ± 1° (c=2, Chloroform)
Formula Weight 364.37
Percent Purity 99%
Chemical Name or Material 1-Thio-beta-D-glucose tetraacetate
EINECSNumber 243-392-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only