accessibility menu, dialog, popup

UserName

CAS RN 19879-84-6

IUPAC Name: [3,4,5-tris(acetyloxy)-6-sulfanyloxan-2-yl]methyl acetate
Synonyms: [3,4,5-tris(acetyloxy)-6-sulfanyloxan-2-yl]methyl acetate
Molecular Weight (g/mol): 364.37
Molecular Formula: C14H20O9S
InChi Key: SFOZKJGZNOBSHF-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(S)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results