missing translation for 'onlineSavingsMsg'
Learn More

1-Thieno[3,2-d]pyrimidin-4-ylpiperidine-4-carbaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 916766-91-1
Molecular Formula C12H13N3OS
Molecular Weight (g/mol) 247.316
MDL Number MFCD09817540
InChI Key YGMBYHWGCCTCNN-UHFFFAOYSA-N
Synonym 1-thieno 3,2-d pyrimidin-4-ylpiperidine-4-carbaldehyde, 1-thieno 3,2-d pyrimidin-4-yl piperidine-4-carboxaldehyde, 1-thieno 3,2-d pyrimidin-4-yl piperidine-4-carbaldehyde, 4-4-formylpiperidin-1-yl thieno 3,2-d pyrimidine, 4-formyl-1-thieno 3,2-d pyrimidin-4-yl piperidine, 1-thiopheno 2,3-e pyrimidin-4-ylpiperidine-4-carbaldehyde
PubChem CID 24229706
IUPAC Name 1-thieno[3,2-d]pyrimidin-4-ylpiperidine-4-carbaldehyde
SMILES C1CN(CCC1C=O)C2=NC=NC3=C2SC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC63804CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC63804DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

916766-91-1
247.316
YGMBYHWGCCTCNN-UHFFFAOYSA-N
24229706
C1CN(CCC1C=O)C2=NC=NC3=C2SC=C3
C12H13N3OS
MFCD09817540
1-thieno 3,2-d pyrimidin-4-ylpiperidine-4-carbaldehyde, 1-thieno 3,2-d pyrimidin-4-yl piperidine-4-carboxaldehyde, 1-thieno 3,2-d pyrimidin-4-yl piperidine-4-carbaldehyde, 4-4-formylpiperidin-1-yl thieno 3,2-d pyrimidine, 4-formyl-1-thieno 3,2-d pyrimidin-4-yl piperidine, 1-thiopheno 2,3-e pyrimidin-4-ylpiperidine-4-carbaldehyde
1-thieno[3,2-d]pyrimidin-4-ylpiperidine-4-carbaldehyde

Specifications

916766-91-1
100.0
C12H13N3OS
1-thieno 3,2-d pyrimidin-4-ylpiperidine-4-carbaldehyde, 1-thieno 3,2-d pyrimidin-4-yl piperidine-4-carboxaldehyde, 1-thieno 3,2-d pyrimidin-4-yl piperidine-4-carbaldehyde, 4-4-formylpiperidin-1-yl thieno 3,2-d pyrimidine, 4-formyl-1-thieno 3,2-d pyrimidin-4-yl piperidine, 1-thiopheno 2,3-e pyrimidin-4-ylpiperidine-4-carbaldehyde
C1CN(CCC1C=O)C2=NC=NC3=C2SC=C3
247.316
247.32
1-Thieno[3, 2-d]pyrimidin-4-ylpiperidine-4-carbaldehyde
96.5
97%
MFCD09817540
YGMBYHWGCCTCNN-UHFFFAOYSA-N
1-thieno[3,2-d]pyrimidin-4-ylpiperidine-4-carbaldehyde
24229706
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents