missing translation for 'onlineSavingsMsg'
Learn More

1-Thieno[3,2-d]pyrimidin-4-ylpiperidine-4-carbaldehyde, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC63804DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC63804DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

916766-91-1
247.316
YGMBYHWGCCTCNN-UHFFFAOYSA-N
24229706
C1CN(CCC1C=O)C2=NC=NC3=C2SC=C3
C12H13N3OS
MFCD09817540
1-thieno 3,2-d pyrimidin-4-ylpiperidine-4-carbaldehyde, 1-thieno 3,2-d pyrimidin-4-yl piperidine-4-carboxaldehyde, 1-thieno 3,2-d pyrimidin-4-yl piperidine-4-carbaldehyde, 4-4-formylpiperidin-1-yl thieno 3,2-d pyrimidine, 4-formyl-1-thieno 3,2-d pyrimidin-4-yl piperidine, 1-thiopheno 2,3-e pyrimidin-4-ylpiperidine-4-carbaldehyde
1-thieno[3,2-d]pyrimidin-4-ylpiperidine-4-carbaldehyde

Specifications

1-Thieno[3, 2-d]pyrimidin-4-ylpiperidine-4-carbaldehyde
96.5
97%
C12H13N3OS
MFCD09817540
YGMBYHWGCCTCNN-UHFFFAOYSA-N
1-thieno[3,2-d]pyrimidin-4-ylpiperidine-4-carbaldehyde
24229706
97%
916766-91-1
100.0
Amber Glass Bottle
1g
1-thieno 3,2-d pyrimidin-4-ylpiperidine-4-carbaldehyde, 1-thieno 3,2-d pyrimidin-4-yl piperidine-4-carboxaldehyde, 1-thieno 3,2-d pyrimidin-4-yl piperidine-4-carbaldehyde, 4-4-formylpiperidin-1-yl thieno 3,2-d pyrimidine, 4-formyl-1-thieno 3,2-d pyrimidin-4-yl piperidine, 1-thiopheno 2,3-e pyrimidin-4-ylpiperidine-4-carbaldehyde
C1CN(CCC1C=O)C2=NC=NC3=C2SC=C3
247.316
247.32
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions