missing translation for 'onlineSavingsMsg'
Learn More

(1-Thieno[3,2-d]pyrimidin-4-ylpiperid-4-yl)methylamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 937795-94-3
Molecular Formula C12H16N4S
Molecular Weight (g/mol) 248.35
MDL Number MFCD09787494
InChI Key ZKNFKYHZGAWSEG-UHFFFAOYSA-N
Synonym 1-thieno 3,2-d pyrimidin-4-ylpiperid-4-yl methylamine, 1-thieno 3,2-d pyrimidin-4-yl piperidin-4-yl methanamine, 1-1-thieno 3,2-d pyrimidin-4-yl piperidin-4-yl methanamine, 1-thiopheno 2,3-e pyrimidin-4-yl-4-piperidyl methylamine, 4-4-aminomethyl piperidin-1-yl thieno 3,2-d pyrimidine
PubChem CID 24229708
IUPAC Name 1-(1-{thieno[3,2-d]pyrimidin-4-yl}piperidin-4-yl)methanamine
SMILES NCC1CCN(CC1)C1=C2SC=CC2=NC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC63813CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC63813DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

937795-94-3
248.35
ZKNFKYHZGAWSEG-UHFFFAOYSA-N
24229708
NCC1CCN(CC1)C1=C2SC=CC2=NC=N1
C12H16N4S
MFCD09787494
1-thieno 3,2-d pyrimidin-4-ylpiperid-4-yl methylamine, 1-thieno 3,2-d pyrimidin-4-yl piperidin-4-yl methanamine, 1-1-thieno 3,2-d pyrimidin-4-yl piperidin-4-yl methanamine, 1-thiopheno 2,3-e pyrimidin-4-yl-4-piperidyl methylamine, 4-4-aminomethyl piperidin-1-yl thieno 3,2-d pyrimidine
1-(1-{thieno[3,2-d]pyrimidin-4-yl}piperidin-4-yl)methanamine

Specifications

937795-94-3
100.0
C12H16N4S
1-thieno 3,2-d pyrimidin-4-ylpiperid-4-yl methylamine, 1-thieno 3,2-d pyrimidin-4-yl piperidin-4-yl methanamine, 1-1-thieno 3,2-d pyrimidin-4-yl piperidin-4-yl methanamine, 1-thiopheno 2,3-e pyrimidin-4-yl-4-piperidyl methylamine, 4-4-aminomethyl piperidin-1-yl thieno 3,2-d pyrimidine
NCC1CCN(CC1)C1=C2SC=CC2=NC=N1
248.35
248.35
(1-Thieno[3, 2-d]pyrimidin-4-ylpiperid-4-yl)methylamine
96.5
97%
MFCD09787494
ZKNFKYHZGAWSEG-UHFFFAOYSA-N
1-(1-{thieno[3,2-d]pyrimidin-4-yl}piperidin-4-yl)methanamine
24229708
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents