missing translation for 'onlineSavingsMsg'
Learn More

(1-Thieno[3,2-d]pyrimidin-4-ylpiperid-4-yl)methylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC63813CB

Quantity 250mg
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC63813CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

937795-94-3
248.35
ZKNFKYHZGAWSEG-UHFFFAOYSA-N
24229708
NCC1CCN(CC1)C1=C2SC=CC2=NC=N1
C12H16N4S
MFCD09787494
1-thieno 3,2-d pyrimidin-4-ylpiperid-4-yl methylamine, 1-thieno 3,2-d pyrimidin-4-yl piperidin-4-yl methanamine, 1-1-thieno 3,2-d pyrimidin-4-yl piperidin-4-yl methanamine, 1-thiopheno 2,3-e pyrimidin-4-yl-4-piperidyl methylamine, 4-4-aminomethyl piperidin-1-yl thieno 3,2-d pyrimidine
1-(1-{thieno[3,2-d]pyrimidin-4-yl}piperidin-4-yl)methanamine

Specifications

(1-Thieno[3, 2-d]pyrimidin-4-ylpiperid-4-yl)methylamine
96.5
97%
C12H16N4S
MFCD09787494
ZKNFKYHZGAWSEG-UHFFFAOYSA-N
1-(1-{thieno[3,2-d]pyrimidin-4-yl}piperidin-4-yl)methanamine
24229708
97%
937795-94-3
100.0
Amber Glass Bottle
250mg
1-thieno 3,2-d pyrimidin-4-ylpiperid-4-yl methylamine, 1-thieno 3,2-d pyrimidin-4-yl piperidin-4-yl methanamine, 1-1-thieno 3,2-d pyrimidin-4-yl piperidin-4-yl methanamine, 1-thiopheno 2,3-e pyrimidin-4-yl-4-piperidyl methylamine, 4-4-aminomethyl piperidin-1-yl thieno 3,2-d pyrimidine
NCC1CCN(CC1)C1=C2SC=CC2=NC=N1
248.35
248.35
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents