missing translation for 'onlineSavingsMsg'
Learn More

[1-(Thien-2-ylmethyl)piperid-4-yl]methanol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 926921-81-5
Molecular Formula C11H17NOS
Molecular Weight (g/mol) 211.323
MDL Number MFCD09817554
InChI Key KAZWOHUGLHRSDG-UHFFFAOYSA-N
Synonym 1-thien-2-ylmethyl piperid-4-yl methanol, 1-thien-2-ylmethyl piperidin-4-yl methanol, 1-thiophen-2-ylmethyl piperidin-4-yl methanol, 1-2-thienylmethyl-4-piperidyl methan-1-ol, 1-thiophen-2-yl methyl piperidin-4-yl methanol, 4-piperidinemethanol, 1-2-thienylmethyl-, hydrochloride
PubChem CID 24229736
IUPAC Name [1-(thiophen-2-ylmethyl)piperidin-4-yl]methanol
SMILES C1CN(CCC1CO)CC2=CC=CS2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC66709CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC66709DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

926921-81-5
211.323
KAZWOHUGLHRSDG-UHFFFAOYSA-N
24229736
C1CN(CCC1CO)CC2=CC=CS2
C11H17NOS
MFCD09817554
1-thien-2-ylmethyl piperid-4-yl methanol, 1-thien-2-ylmethyl piperidin-4-yl methanol, 1-thiophen-2-ylmethyl piperidin-4-yl methanol, 1-2-thienylmethyl-4-piperidyl methan-1-ol, 1-thiophen-2-yl methyl piperidin-4-yl methanol, 4-piperidinemethanol, 1-2-thienylmethyl-, hydrochloride
[1-(thiophen-2-ylmethyl)piperidin-4-yl]methanol

Specifications

926921-81-5
100.0
C11H17NOS
1-thien-2-ylmethyl piperid-4-yl methanol, 1-thien-2-ylmethyl piperidin-4-yl methanol, 1-thiophen-2-ylmethyl piperidin-4-yl methanol, 1-2-thienylmethyl-4-piperidyl methan-1-ol, 1-thiophen-2-yl methyl piperidin-4-yl methanol, 4-piperidinemethanol, 1-2-thienylmethyl-, hydrochloride
C1CN(CCC1CO)CC2=CC=CS2
211.323
211.33
[1-(Thien-2-ylmethyl)piperid-4-yl]methanol
96.5
97%
MFCD09817554
KAZWOHUGLHRSDG-UHFFFAOYSA-N
[1-(thiophen-2-ylmethyl)piperidin-4-yl]methanol
24229736
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents