missing translation for 'onlineSavingsMsg'
Learn More

[1-(Thien-2-ylmethyl)piperid-4-yl]methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC66709DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC66709DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

926921-81-5
211.323
KAZWOHUGLHRSDG-UHFFFAOYSA-N
24229736
C1CN(CCC1CO)CC2=CC=CS2
C11H17NOS
MFCD09817554
1-thien-2-ylmethyl piperid-4-yl methanol, 1-thien-2-ylmethyl piperidin-4-yl methanol, 1-thiophen-2-ylmethyl piperidin-4-yl methanol, 1-2-thienylmethyl-4-piperidyl methan-1-ol, 1-thiophen-2-yl methyl piperidin-4-yl methanol, 4-piperidinemethanol, 1-2-thienylmethyl-, hydrochloride
[1-(thiophen-2-ylmethyl)piperidin-4-yl]methanol

Specifications

[1-(Thien-2-ylmethyl)piperid-4-yl]methanol
96.5
97%
C11H17NOS
MFCD09817554
KAZWOHUGLHRSDG-UHFFFAOYSA-N
[1-(thiophen-2-ylmethyl)piperidin-4-yl]methanol
24229736
97%
926921-81-5
100.0
Amber Glass Bottle
1g
1-thien-2-ylmethyl piperid-4-yl methanol, 1-thien-2-ylmethyl piperidin-4-yl methanol, 1-thiophen-2-ylmethyl piperidin-4-yl methanol, 1-2-thienylmethyl-4-piperidyl methan-1-ol, 1-thiophen-2-yl methyl piperidin-4-yl methanol, 4-piperidinemethanol, 1-2-thienylmethyl-, hydrochloride
C1CN(CCC1CO)CC2=CC=CS2
211.323
211.33
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents