missing translation for 'onlineSavingsMsg'
Learn More

1-Tetralone, 97%

Catalog No. AAA1330336
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1330336 500 g
AAA1330322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1330336 Supplier Thermo Scientific Chemicals Supplier No. A1330336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 529-34-0 | C10H10O | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 529-34-0
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00001688
InChI Key XHLHPRDBBAGVEG-UHFFFAOYSA-N
Synonym alpha-tetralone, 1-tetralone, 3,4-dihydronaphthalen-1 2h-one, 1,2,3,4-tetrahydronaphthalen-1-one, 3,4-dihydro-1 2h-naphthalenone, tetralone, a-tetralone, 1-oxotetralin, 1 2h-naphthalenone, 3,4-dihydro, .alpha.-tetralone
PubChem CID 10724
IUPAC Name 3,4-dihydro-2H-naphthalen-1-one
SMILES O=C1CCCC2=CC=CC=C12

Specifications

Melting Point 6°C to 8°C
Density 1.097
Boiling Point 257°C
Flash Point 129°C (264°F)
Refractive Index 1.5695
Quantity 500 g
Beilstein 607374
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material 1-Tetralone
Hazard Category H302-H331
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P311-P330-P501c
EINECSNumber 208-460-6
RTECSNumber QK4375000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only