missing translation for 'onlineSavingsMsg'
Learn More

1-Tetrahydropyran-4-yl-1H-pyrazol-4-ylamine, 97%

Catalog No. p-7077422
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453570010 1 g
AC453570050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453570010 Supplier Thermo Scientific Chemicals Supplier No. 453570010

CAS: 1190380-49-4 | C8H13N3O | 167.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1190380-49-4
Molecular Formula C8H13N3O
Molecular Weight (g/mol) 167.21
MDL Number MFCD11505007
InChI Key GZZNBQLBGSVOOZ-UHFFFAOYSA-N
Synonym 1-tetrahydro-2h-pyran-4-yl-1h-pyrazol-4-amine, 1-oxan-4-yl pyrazol-4-amine, 1-oxan-4-yl-1h-pyrazol-4-amine, 1h-pyrazol-4-amine, 1-tetrahydro-2h-pyran-4-yl, 1-tetrahydropyran-4-yl pyrazol-4-amine, 1-tetrahydro-pyran-4-yl-1h-pyrazol-4-ylamine, 1-2h-3,4,5,6-tetrahydropyran-4-yl pyrazole-4-ylamine
PubChem CID 51063703
SMILES NC1=CN(N=C1)C1CCOCC1

Specifications

Melting Point 105.0°C to 107.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
IUPAC Name 1-(oxan-4-yl)-1H-pyrazol-4-amine
Formula Weight 167.21
Percent Purity 97%
Chemical Name or Material 1-Tetrahydropyran-4-yl-1H-pyrazol-4-ylamine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only