missing translation for 'onlineSavingsMsg'
Learn More

1-(Tetrahydropyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole 98.0+%, TCI America™

Catalog No. p-7137640
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32151G 1 g
T32155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. T32151G missing translation for 'mfr' TCI America missing translation for 'supplierNo' T32151G

Chemical Identifiers

CAS 903550-26-5
Molecular Formula C14H23BN2O3
Molecular Weight (g/mol) 278.159
MDL Number MFCD09037501
InChI Key ZZRFDLHBMBHJTI-UHFFFAOYSA-N
Synonym 1-tetrahydro-2h-pyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-2-tetrahydropyranyl-1h-pyrazole-5-boronic acid pinacol ester, 1-tetrahydro-2h-pyran-2-yl-1h-pyrazole-5-boronic acid pinacol ester, 1-tetrahydropyran-2-yl-1h-pyrazole-5-boronic acid pinacol ester, 1-oxan-2-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-oxan-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-oxan-2-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-oxan-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-y-1h-pyrazole, 1-tetrahydropyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole
PubChem CID 11587208
IUPAC Name 1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=NN2C3CCCCO3

Specifications

Melting Point 76°C
Color White
Quantity 1 g
Formula Weight 278.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Tetrahydropyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
TSCA No
Recommended Storage Refrigerator