accessibility menu, dialog, popup

UserName

CAS RN 903550-26-5

IUPAC Name: 1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol): 278.16
Molecular Formula: C14H23BN2O3
InChi Key: ZZRFDLHBMBHJTI-UHFFFAOYNA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=NN1C1CCCCO1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)