missing translation for 'onlineSavingsMsg'
Learn More

1-(Tetrahydro-2H-pyran-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 98.0+%, TCI America™

Catalog No. T3668200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3668200MG 200 mg
T36681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3668200MG Supplier TCI America Supplier No. T3668200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1040377-03-4
Molecular Formula C14H23BN2O3
Molecular Weight (g/mol) 278.159
MDL Number MFCD12033229
InChI Key BOOVIFJKQGYEON-UHFFFAOYSA-N
Synonym 1-(Tetrahydro-2H-pyran-4-yl)-1H-pyrazole-4-boronic Acid Pinacol Ester
PubChem CID 45788496
IUPAC Name 1-(oxan-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C3CCOCC3

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 200 mg
Formula Weight 278.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Tetrahydro-2H-pyran-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole