accessibility menu, dialog, popup

UserName

CAS RN 1040377-03-4

IUPAC Name: 1-(oxan-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 1-(oxan-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol): 278.16
Molecular Formula: C14H23BN2O3
InChi Key: BOOVIFJKQGYEON-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN(N=C1)C1CCOCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results