missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Tetrahydro-2-furoyl)piperazine 97.0+%, TCI America™
$70.84 - $186.41
Chemical Identifiers
| CAS | 63074-07-7 |
|---|---|
| Molecular Formula | C9H16N2O2 |
| Molecular Weight (g/mol) | 184.239 |
| MDL Number | MFCD00190247 |
| InChI Key | UKESBLFBQANJHH-UHFFFAOYSA-N |
| Synonym | 1-tetrahydro-2-furoyl piperazine, 1-2-tetrahydrofuroyl piperazine, piperazin-1-yl tetrahydrofuran-2-yl methanone, 1-2-tetrahydrofuroyl-piperazine, n-2-tetrahydrofuroyl piperazine, 1-oxolane-2-carbonyl piperazine, 1-tetrahydrofuran-2-ylcarbonyl piperazine, n-tetrahydro-2-furoyl piperazine, 1-oxolan-2-ylcarbonyl piperazine, oxolan-2-yl piperazin-1-yl methanone |
| PubChem CID | 2734641 |
| IUPAC Name | oxolan-2-yl(piperazin-1-yl)methanone |
| SMILES | C1CC(OC1)C(=O)N2CCNCC2 |
Chemical Identifiers
| 63074-07-7 | |
| 184.239 | |
| UKESBLFBQANJHH-UHFFFAOYSA-N | |
| 2734641 | |
| C1CC(OC1)C(=O)N2CCNCC2 |
| C9H16N2O2 | |
| MFCD00190247 | |
| 1-tetrahydro-2-furoyl piperazine, 1-2-tetrahydrofuroyl piperazine, piperazin-1-yl tetrahydrofuran-2-yl methanone, 1-2-tetrahydrofuroyl-piperazine, n-2-tetrahydrofuroyl piperazine, 1-oxolane-2-carbonyl piperazine, 1-tetrahydrofuran-2-ylcarbonyl piperazine, n-tetrahydro-2-furoyl piperazine, 1-oxolan-2-ylcarbonyl piperazine, oxolan-2-yl piperazin-1-yl methanone | |
| oxolan-2-yl(piperazin-1-yl)methanone |
Specifications
| 63074-07-7 | |
| 5 g | |
| C9H16N2O2 | |
| 1-tetrahydro-2-furoyl piperazine, 1-2-tetrahydrofuroyl piperazine, piperazin-1-yl tetrahydrofuran-2-yl methanone, 1-2-tetrahydrofuroyl-piperazine, n-2-tetrahydrofuroyl piperazine, 1-oxolane-2-carbonyl piperazine, 1-tetrahydrofuran-2-ylcarbonyl piperazine, n-tetrahydro-2-furoyl piperazine, 1-oxolan-2-ylcarbonyl piperazine, oxolan-2-yl piperazin-1-yl methanone | |
| C1CC(OC1)C(=O)N2CCNCC2 | |
| 184.239 | |
| 184.24 | |
| 1-(Tetrahydro-2-furoyl)piperazine |
| 125°C | |
| Clear Liquid at 20°C | |
| MFCD00190247 | |
| UKESBLFBQANJHH-UHFFFAOYSA-N | |
| oxolan-2-yl(piperazin-1-yl)methanone | |
| 2734641 | |
| ≥97.0% (GC) |
Safety and Handling
TSCA : No