accessibility menu, dialog, popup

UserName

O(S)ONNH2

CAS RN 63074-07-7

IUPAC Name: 4-[(2S)-oxolane-2-carbonyl]piperazin-1-ium
Synonyms: 4-[(2S)-oxolane-2-carbonyl]piperazin-1-ium
Molecular Weight (g/mol): 185.25
Molecular Formula: C9H17N2O2
InChi Key: UKESBLFBQANJHH-QMMMGPOBSA-O
SMILES: O=C([C@@H]1CCCO1)N1CC[NH2+]CC1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)